Assay Details
| Gene Symbol: Ebpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500499.2 | XP_003500547.1 | 3 - 4 | 422 | 80 | |
| GenBank mRNA | JI885879.1 | - | 3 - 4 | 233 | 80 | |
| JP039303.1 | - | 3 - 4 | 233 | 80 | ||
More Information
Set Membership: |
|

Ebpl
Genomic Map