Assay Details
Gene Symbol: Ccdc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502669.2 | XP_003502717.1 | 2 - 3 | 337 | 62 | |
XM_007643302.1 | XP_007641492.1 | 3 - 4 | 378 | 62 | ||
XM_007643303.1 | XP_007641493.1 | 2 - 3 | 284 | 62 | ||
GenBank mRNA | JI896332.1 | - | 1 - 2 | 147 | 62 | |
JP045447.1 | - | 1 - 2 | 147 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
