Assay Details
| Gene Symbol: Lgals4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503817.1 | XP_003503865.1 | 1 - 2 | 45 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lgals4| Gene Symbol: Lgals4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503817.1 | XP_003503865.1 | 1 - 2 | 45 | 58 | |
Set Membership: |
|