Assay Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504510.2 | XP_003504558.1 | 3 - 4 | 613 | 60 | |
| XM_007645057.1 | XP_007643247.1 | 3 - 4 | 613 | 60 | ||
More Information
Set Membership: |
|

Gfap
Genomic Map