Assay Details
| Gene Symbol: Mkrn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507344.2 | XP_003507392.1 | 3 - 4 | 624 | 72 | |
| GenBank mRNA | JI875487.1 | - | 3 - 4 | 625 | 72 | |
| JP053910.1 | - | 3 - 4 | 625 | 72 | ||
More Information
Set Membership: |
|

Mkrn2
Genomic Map