Assay Details
| Gene Symbol: Pidd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509765.2 | XP_003509813.1 | 5 - 6 | 1211 | 81 | |
| XM_007650419.1 | XP_007648609.1 | 5 - 6 | 1116 | 81 | ||
| XM_007650420.1 | XP_007648610.1 | 5 - 6 | 1207 | 81 | ||
| XM_007650421.1 | XP_007648611.1 | 5 - 6 | 1255 | 81 | ||
More Information
Set Membership: |
|

Pidd
Genomic Map