Assay Details
| Gene Symbol: Pidd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509765.2 | XP_003509813.1 | 7 - 8 | 1543 | 78 | |
| XM_007650419.1 | XP_007648609.1 | 7 - 8 | 1448 | 78 | ||
| XM_007650420.1 | XP_007648610.1 | 7 - 8 | 1539 | 78 | ||
| XM_007650421.1 | XP_007648611.1 | 7 - 8 | 1587 | 78 | ||
| GenBank mRNA | JI865071.1 | - | 5 - 6 | 1254 | 78 | |
| JP040891.1 | - | 5 - 6 | 1254 | 78 | ||
More Information
Set Membership: |
|

Pidd
Genomic Map