Assay Details
| Gene Symbol: Pidd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509765.2 | XP_003509813.1 | 9 - 10 | 1867 | 87 | |
| XM_007650419.1 | XP_007648609.1 | 9 - 10 | 1772 | 87 | ||
| XM_007650420.1 | XP_007648610.1 | 9 - 10 | 1863 | 87 | ||
| XM_007650421.1 | XP_007648611.1 | 9 - 10 | 1911 | 87 | ||
More Information
Set Membership: |
|

Pidd
Genomic Map