Assay Details
| Gene Symbol: LOC100768178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511519.1 | XP_003511567.1 | 1 | 221 | 74 | |
| XM_007652069.1 | XP_007650259.1 | 1 | 221 | 74 | ||
More Information
Set Membership: |
|

LOC100768178
Genomic Map