Assay Details
| Gene Symbol: Atp6ap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512282.2 | XP_003512330.2 | 3 - 4 | 587 | 61 | |
| GenBank mRNA | JI880606.1 | - | 3 - 4 | 587 | 61 | |
| JP054439.1 | - | 3 - 4 | 587 | 61 | ||
More Information
Set Membership: |
|

Atp6ap1
Genomic Map