Assay Details
| Gene Symbol: LOC100768178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652070.1 | XP_007650260.1 | 1 - 2 | 115 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100768178| Gene Symbol: LOC100768178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007652070.1 | XP_007650260.1 | 1 - 2 | 115 | 55 | |
Set Membership: |
|