Assay Details
| Gene Symbol: Dusp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645035.1 | XP_007643225.1 | 3 - 4 | 411 | 53 | |
| GenBank mRNA | JI864761.1 | - | 3 - 4 | 416 | 53 | |
More Information
Set Membership: |
|

Dusp3
Genomic Map