Assay Details
Gene Symbol: Znf354a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498495.2 | XP_003498543.1 | 4 - 5 | 438 | 85 | |
GenBank mRNA | JI889380.1 | - | 4 - 5 | 376 | 85 | |
JP050625.1 | - | 4 - 5 | 376 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
