Assay Details
| Gene Symbol: Mrpl35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003503890.2 | XP_003503938.1 | 2 - 3 | 316 | 67 | |
| GenBank mRNA | JI879363.1 | - | 2 - 3 | 308 | 67 | |
More Information
Set Membership: |
|

Mrpl35
Genomic Map