Assay Details
| Gene Symbol: Dydc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505660.1 | XP_003505708.1 | 1 - 2 | 149 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dydc2| Gene Symbol: Dydc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505660.1 | XP_003505708.1 | 1 - 2 | 149 | 93 | |
Set Membership: |
|