Assay Details
| Gene Symbol: Nip7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508682.2 | XP_003508730.1 | 2 - 3 | 207 | 57 | |
| GenBank mRNA | JI872140.1 | - | 2 - 3 | 202 | 57 | |
More Information
Set Membership: |
|

Nip7
Genomic Map