Assay Details
| Gene Symbol: Psph | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509485.2 | XP_003509533.1 | 3 - 4 | 515 | 69 | |
| GenBank mRNA | JP063064.1 | - | 3 - 4 | 532 | 69 | |
More Information
Set Membership: |
|

Psph
Genomic Map