Assay Details
| Gene Symbol: LOC100770124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007613063.1 | XP_007611253.1 | 2 - 3 | 317 | 86 | |
| GenBank mRNA | JI894026.1 | - | 1 - 2 | 168 | 86 | |
More Information
Set Membership: |
|

LOC100770124
Genomic Map