Assay Details
| Gene Symbol: Tbca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007632011.1 | XP_007630201.1 | 2 - 3 | 126 | 71 | |
| XM_007632020.1 | XP_007630210.1 | 2 - 3 | 263 | 71 | ||
| GenBank mRNA | JP039729.1 | - | 1 - 2 | 198 | 71 | |
More Information
Set Membership: |
|

Tbca
Genomic Map