Assay Details
| Gene Symbol: LOC100751702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497001.2 | XP_003497049.2 | 24 - 25 | 3219 | 83 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100751702| Gene Symbol: LOC100751702 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497001.2 | XP_003497049.2 | 24 - 25 | 3219 | 83 | |