Assay Details
| Gene Symbol: Ube2e2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498207.2 | XP_003498255.2 | 2 - 3 | 286 | 64 | |
| GenBank mRNA | JI864665.1 | - | 2 - 3 | 286 | 64 | |
More Information
Set Membership: |
|

Ube2e2
Genomic Map