Assay Details
| Gene Symbol: Rfxank | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003499110.2 | XP_003499158.2 | 6 - 7 | 629 | 61 | |
| XM_007637614.1 | XP_007635804.1 | 7 - 8 | 731 | 61 | ||
| XM_007637620.1 | XP_007635810.1 | 7 - 8 | 1028 | 61 | ||
| XM_007637633.1 | XP_007635823.1 | 5 - 6 | 464 | 61 | ||
| XM_007637641.1 | XP_007635831.1 | 5 - 6 | 708 | 61 | ||
| XM_007637647.1 | XP_007635837.1 | 5 - 6 | 485 | 61 | ||
| GenBank mRNA | JI893741.1 | - | 2 - 3 | 61 | ||
More Information
Set Membership: |
|

Rfxank
Genomic Map