Assay Details
| Gene Symbol: Blmh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506081.2 | XP_003506129.1 | 9 - 10 | 1192 | 63 | |
| GenBank mRNA | JI876434.1 | - | 9 - 10 | 1174 | 63 | |
| JP056842.1 | - | 9 - 10 | 1174 | 63 | ||
More Information
Set Membership: |
|

Blmh
Genomic Map