Assay Details
| Gene Symbol: Blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003508272.2 | XP_003508320.1 | 3 - 4 | 883 | 69 | |
| GenBank mRNA | JI864094.1 | - | 3 - 4 | 691 | 69 | |
| JP049992.1 | - | 3 - 4 | 691 | 69 | ||
More Information
Set Membership: |
|

Blzf1
Genomic Map