Assay Details
| Gene Symbol: Zkscan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510414.2 | XP_003510462.1 | 2 - 3 | 633 | 84 | |
| XM_007651033.1 | XP_007649223.1 | 2 - 3 | 633 | 84 | ||
More Information
Set Membership: |
|

Zkscan1
Genomic Map