Assay Details
| Gene Symbol: Cript | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003511401.2 | XP_003511449.1 | 4 - 5 | 314 | 81 | |
| GenBank mRNA | JI889249.1 | - | 4 - 5 | 307 | 81 | |
| JP058223.1 | - | 4 - 5 | 307 | 81 | ||
More Information
Set Membership: |
|

Cript
Genomic Map