Assay Details
Gene Symbol: LOC100774451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007620022.1 | XP_007618212.1 | 3 - 4 | 2542 | 93 | |
XM_007620030.1 | XP_007618220.1 | 3 - 4 | 2273 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
