Assay Details
| Gene Symbol: Sla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497089.2 | XP_003497137.1 | 3 - 4 | 283 | 60 | |
| XM_007607743.1 | XP_007605933.1 | 3 - 4 | 283 | 60 | ||
More Information
Set Membership: |
|

Sla2
Genomic Map