Assay Details
| Gene Symbol: Rtfdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003501438.2 | XP_003501486.1 | 4 - 5 | 471 | 55 | |
| XM_007642097.1 | XP_007640287.1 | 4 - 5 | 471 | 55 | ||
| GenBank mRNA | JI876115.1 | - | 4 - 5 | 469 | 55 | |
| JP046996.1 | - | 4 - 5 | 469 | 55 | ||
More Information
Set Membership: |
|

Rtfdc1
Genomic Map