Assay Details
| Gene Symbol: Mrpl40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502445.2 | XP_003502493.1 | 1 - 2 | 207 | 86 | |
| GenBank mRNA | JI882302.1 | - | 1 - 2 | 114 | 86 | |
| JP040152.1 | - | 1 - 2 | 114 | 86 | ||
More Information
Set Membership: |
|

Mrpl40
Genomic Map