Assay Details
| Gene Symbol: Znf76 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506290.2 | XP_003506338.1 | 4 - 5 | 541 | 56 | |
| XM_007646796.1 | XP_007644986.1 | 4 - 5 | 542 | 56 | ||
| GenBank mRNA | JI877658.1 | - | 4 - 5 | 466 | 56 | |
| JP051495.1 | - | 4 - 5 | 466 | 56 | ||
More Information
Set Membership: |
|

Znf76
Genomic Map