Assay Details
| Gene Symbol: Ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003506307.2 | XP_003506355.1 | 3 - 4 | 582 | 70 | |
| GenBank mRNA | JI881623.1 | - | 3 - 4 | 444 | 70 | |
| JP052603.1 | - | 3 - 4 | 444 | 70 | ||
More Information
Set Membership: |
|

Ak3
Genomic Map