Assay Details
| Gene Symbol: Pet100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507745.2 | XP_003507793.1 | 3 - 4 | 183 | 69 | |
| XM_007648342.1 | XP_007646532.1 | 3 - 4 | 328 | 69 | ||
| XM_007648343.1 | XP_007646533.1 | 3 - 4 | 269 | 69 | ||
| XM_007648344.1 | XP_007646534.1 | 3 - 4 | 110 | 69 | ||
| GenBank mRNA | JI877053.1 | - | 2 - 3 | 76 | 69 | |
More Information
Set Membership: |
|

Pet100
Genomic Map