Assay Details
| Gene Symbol: LOC100765374 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003512348.2 | XP_003512396.1 | 1 - 2 | 202 | 76 | |
| GenBank mRNA | JI893785.1 | - | 1 - 2 | 171 | 76 | |
| JP041971.1 | - | 1 - 2 | 171 | 76 | ||
More Information
Set Membership: |
|

LOC100765374
Genomic Map