Assay Details
| Gene Symbol: Cd151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007650423.1 | XP_007648613.1 | 6 - 7 | 720 | 65 | |
| XM_007650424.1 | XP_007648614.1 | 5 - 6 | 689 | 65 | ||
| GenBank mRNA | JI891745.1 | - | 1 - 2 | 163 | 65 | |
More Information
Set Membership: |
|

Cd151
Genomic Map