Assay Details
| Gene Symbol: Plcxd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003497845.2 | XP_003497893.1 | 2 - 3 | 1169 | 62 | |
| XM_007618573.1 | XP_007616763.1 | 2 - 3 | 1170 | 62 | ||
| XM_007618579.1 | XP_007616769.1 | 2 - 3 | 1169 | 62 | ||
More Information
Set Membership: |
|

Plcxd2
Genomic Map