Assay Details
| Gene Symbol: LOC100771756 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498089.2 | XP_003498137.1 | 18 - 19 | 1973 | 77 | |
| GenBank mRNA | JI890269.1 | - | 5 - 6 | 596 | 77 | |
| JP053420.1 | - | 5 - 6 | 596 | 77 | ||
More Information
Set Membership: |
|

LOC100771756
Genomic Map