Assay Details
| Gene Symbol: Pcgf5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007636034.1 | XP_007634224.1 | 1 - 2 | 135 | 71 | |
| XM_007636041.1 | XP_007634231.1 | 1 - 2 | 135 | 71 | ||
| XM_007636048.1 | XP_007634238.1 | 1 - 2 | 268 | 71 | ||
| GenBank mRNA | JP047728.1 | - | 1 - 2 | 344 | 71 | |
More Information
Set Membership: |
|

Pcgf5
Genomic Map