Assay Details
| Gene Symbol: LOC100762989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007616046.1 | XP_007614236.1 | 4 - 5 | 737 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100762989| Gene Symbol: LOC100762989 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007616046.1 | XP_007614236.1 | 4 - 5 | 737 | 147 | |
Set Membership: |
|