Assay Details
| Gene Symbol: Ube2d2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647874.1 | XP_007646064.1 | 2 - 3 | 126 | 89 | |
| GenBank mRNA | JP062965.1 | - | 2 - 3 | 126 | 89 | |
More Information
Set Membership: |
|

Ube2d2
Genomic Map