Assay Details
| Gene Symbol: Camp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644791.1 | XP_007642981.1 | 5 - 6 | 1024 | 104 | |
| XM_007644792.1 | XP_007642982.1 | 4 - 5 | 905 | 104 | ||
| XM_007644793.1 | XP_007642983.1 | 3 - 4 | 841 | 104 | ||
| XM_007644794.1 | XP_007642984.1 | 4 - 5 | 960 | 104 | ||
More Information
Set Membership: |
|

Camp
Genomic Map