Assay Details
| Gene Symbol: Calm1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007642064.1 | XP_007640254.1 | 4 - 5 | 606 | 75 | |
| GenBank mRNA | JI886049.1 | - | 4 - 5 | 606 | 75 | |
More Information
Set Membership: |
|

Calm1
Genomic Map