Assay Details
| Gene Symbol: LOC100756919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500483.2 | XP_003500531.2 | 1 - 2 | 423 | 63 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100756919| Gene Symbol: LOC100756919 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003500483.2 | XP_003500531.2 | 1 - 2 | 423 | 63 | |
Set Membership: | 
		   
					
						
							 |