Assay Details
| Gene Symbol: Dhrs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502049.2 | XP_003502097.1 | 7 - 8 | 1024 | 69 | |
| GenBank mRNA | JI873855.1 | - | 7 - 8 | 928 | 69 | |
| JP041891.1 | - | 7 - 8 | 928 | 69 | ||
More Information
Set Membership: |
|

Dhrs1
Genomic Map