Assay Details
Gene Symbol: Pkib | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504211.2 | XP_003504259.1 | 4 - 5 | 433 | 69 | |
XM_007644746.1 | XP_007642936.1 | 5 - 6 | 556 | 69 | ||
XM_007644747.1 | XP_007642937.1 | 5 - 6 | 463 | 69 | ||
XM_007644748.1 | XP_007642938.1 | 3 - 4 | 371 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
