Assay Details
| Gene Symbol: Suclg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003505635.2 | XP_003505683.1 | 2 - 3 | 238 | 56 | |
| GenBank mRNA | JI879821.1 | - | 2 - 3 | 219 | 56 | |
More Information
Set Membership: |
|

Suclg1
Genomic Map