Assay Details
Gene Symbol: Bend6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003507908.2 | XP_003507956.1 | 2 - 3 | 230 | 71 | |
XM_007648488.1 | XP_007646678.1 | 2 - 3 | 276 | 71 | ||
GenBank mRNA | JI876760.1 | - | 1 - 2 | 95 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
