Assay Details
| Gene Symbol: Cdipt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510118.2 | XP_003510166.1 | 3 - 4 | 715 | 60 | |
| GenBank mRNA | JI867098.1 | - | 4 - 5 | 1051 | 60 | |
| JP037992.1 | - | 3 - 4 | 735 | 60 | ||
More Information
Set Membership: |
|

Cdipt
Genomic Map