Assay Details
| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607817.1 | XP_007606007.1 | 5 - 6 | 795 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sept1| Gene Symbol: Sept1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007607817.1 | XP_007606007.1 | 5 - 6 | 795 | 67 | |
Set Membership: |
|