Assay Details
| Gene Symbol: Nptn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502408.2 | XP_003502456.1 | 4 - 5 | 633 | 62 | |
| GenBank mRNA | JI887235.1 | - | 2 - 3 | 391 | 62 | |
| JP065821.1 | - | 2 - 3 | 390 | 62 | ||
More Information
Set Membership: |
|

Nptn
Genomic Map